missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) oxide, 99%

Catalog No. AAA1788922
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1788922 100 g
AAA1788936 500 g
AAA178890E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1788922 Supplier Thermo Scientific Chemicals Supplier No. A1788922

CAS: 21651-19-4 | OSn | 134.71 g/mol

Tin(II) oxide is used as reducing agent, soft abrasive, and in preparation of stannous salts. It is used in the manufacture of copper ruby glass, and for illumination with UV light.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tin(II) oxide is used as reducing agent, soft abrasive, and in preparation of stannous salts. It is used in the manufacture of copper ruby glass, and for illumination with UV light.

Solubility
Insoluble in water and alcohol.

Notes
Incompatible with materials like strong oxidizing agents, strong acids, strong reducing agents and alkali metals.

Chemical Identifiers

CAS 21651-19-4
Molecular Formula OSn
Molecular Weight (g/mol) 134.71
MDL Number MFCD00011243
InChI Key QHGNHLZPVBIIPX-UHFFFAOYSA-N
Synonym stannous oxide, tin ii oxide, tin oxide sno, tin monoxide, tin oxide sn2o2, stannane, oxo, stannanone, stannicoxide, oxostannanylidene, oxo stannane
PubChem CID 88989
IUPAC Name oxotin
SMILES O=[Sn]

Specifications

Melting Point 1080°C (decomposition)
Quantity 100 g
Linear Formula SnO
Merck Index 14,8787
Solubility Information Insoluble in water and alcohol.
Formula Weight 134.69
Percent Purity 99%
Odor Odorless
Density 6.45
Chemical Name or Material Tin(II) oxide
Hazard Category H335-H373
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P260-P271-P304+P340-P312-P314-P501c
EINECSNumber 244-499-5
RTECSNumber XQ3700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only