missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) oxide, 99.9% (metals basis)

Catalog No. p-4412868
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1156914 25 g
AA1156922 100 g
AA1156936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1156914 Supplier Thermo Scientific Chemicals Supplier No. 01156914

Reducing agent, soft abrasive, preparation of stannous salts | CAS: 21651-19-4 | OSn | 134.71 g/mol

It is used as reducing agent, soft abrasive, and in preparation of stannous salts. It is used in the manufacture of copper ruby glass, and for illumination with UV light.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21651-19-4
Molecular Formula OSn
Molecular Weight (g/mol) 134.71
MDL Number MFCD00011243
InChI Key QHGNHLZPVBIIPX-UHFFFAOYSA-N
Synonym stannous oxide, tin ii oxide, tin oxide sno, tin monoxide, tin oxide sn2o2, stannane, oxo, stannanone, stannicoxide, oxostannanylidene, oxo stannane
PubChem CID 88989
IUPAC Name oxotin
SMILES O=[Sn]

Specifications

Melting Point 1080°C (decomposition)
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Insoluble in water,and alcohol.
Formula Weight 134.69
Density 6.45 g/mL
Chemical Name or Material Tin(II) oxide
Hazard Category H335-H373
Precautionary Statement P260-P271-P304+P340-P312-P314-P501c
EINECSNumber 244-499-5
RTECSNumber XQ3700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only