missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) oxalate, 98%

Catalog No. p-7081640
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1168922 100 g
AAA1168936 500 g
AAA116890E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1168922 Supplier Thermo Scientific Chemicals Supplier No. A1168922
Only null left

CAS: 814-94-8 | C2O4Sn | 206.728 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 814-94-8
Molecular Formula C2O4Sn
Molecular Weight (g/mol) 206.728
MDL Number MFCD00040678
InChI Key OQBLGYCUQGDOOR-UHFFFAOYSA-L
Synonym stannous oxalate, tin ii oxalate, tin oxalate, tin 2+ oxalate, ethanedioic acid, tin 2+ salt 1:1, stavelan cinaty czech, unii-sar72fe8eh, sar72fe8eh, stavelan cinaty
PubChem CID 13149
IUPAC Name oxalate;tin(2+)
SMILES C(=O)(C(=O)[O-])[O-].[Sn+2]

Specifications

Melting Point 280°C (decomposition)
Quantity 100 g
Linear Formula SnC2O4
UN Number UN3288
Beilstein 3708588
Merck Index 14,8786
Formula Weight 206.72
Percent Purity 98%
Density 3.56
Chemical Name or Material Tin (II) oxalate
Hazard Category H302+H312
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 212-414-0
RTECSNumber XQ3950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only