missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) methanesulfonate, 50% w/w aq. soln.

Catalog No. AAH59773AE
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59773AE 100 mL
AAH59773AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59773AE Supplier Thermo Scientific Chemicals Supplier No. H59773AE

CAS: 53408-94-9 | C2H6O6S2Sn | 308.89 g/mol

It is used in electroplating industry. Tin methanesulfonate is an aqueous solution used in the metal surface treatment. Mainly as a replacement for Tin sulfate where high speed deposits are needed (reel-to-reel).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in electroplating industry. Tin methanesulfonate is an aqueous solution used in the metal surface treatment. Mainly as a replacement for Tin sulfate where high speed deposits are needed (reel-to-reel).

Solubility
Not miscible or difficult to mix in water.

Notes
Air sensitive. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 53408-94-9
Molecular Formula C2H6O6S2Sn
Molecular Weight (g/mol) 308.89
MDL Number MFCD00137737
InChI Key JALQQBGHJJURDQ-UHFFFAOYSA-L
Synonym stannous methanesulfonate, methanesulfonic acid, tin 2+ salt, tin ii methanesulfonate, methanesulfonate; tin 2+, methanesulfonic acid, tin 2+ salt 2:1, tin ii methanesulfonate solution, tin ii bis methanesulfonate, pubchem11154, stannous methylsulfonate, 2ch3o3s.sn
PubChem CID 6452824
IUPAC Name methanesulfonate;tin(2+)
SMILES [Sn++].CS([O-])(=O)=O.CS([O-])(=O)=O

Specifications

Melting Point -27°C
Density 1.55
Refractive Index 1.444
Linear Formula (CH3SO3)2Sn
Quantity 100 mL
UN Number UN3265
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 308.91
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Chemical Name or Material Tin(II) methanesulfonate
Show More Show Less
Hazard Category H302-H314-H317-H335
Hazard Statement GHS H Statement
H314-H302-H317
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 401-640-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only