missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) chloride hydrate, Puratronic™, 99.995% (metals basis)

Catalog No. p-7048564
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
5 g
Catalog No. Quantity
AA1089406 5 g
AA1089414 25 g
AA1089422 100 g
3 options

Catalog No. AA1089406

Supplier: Thermo Scientific Chemicals 01089406

Only null left

CAS: 1370709-86-6 | Cl2Sn | 189.61 g/mol

Tin(II) chloride is widely used as a reducing agent in organic synthesis, for example conversion of quinones into hydroquinones, nitroaromatics into aromatic amines, and in Stephen reduction. It can be used to reduce certain metal salts into metals, tin plating, analytical testing of mercury ions, formation of silver mirrors and metal-metal bonds. It is also used for tin-plating, as a mordant in textile dyeing and as a catalyst in the production of polylactic acid. Further it is used as a food additive, wherein it serves as an anti-oxidant and a color retention agent. It can be used in the condensation of aryl aldehydes with cyclohexan-1,3-diones to synthesize xanthenes. It catalyzes direct conversion of aldehydes into beta-keto esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1370709-86-6
Molecular Formula Cl2Sn
Molecular Weight (g/mol) 189.61
MDL Number MFCD00011241
InChI Key AXZWODMDQAVCJE-UHFFFAOYSA-L
Synonym Stannous chloride hydrate
PubChem CID 24479
ChEBI CHEBI:78067
SMILES [Cl-].[Cl-].[Sn++]

Specifications

Melting Point 43°C to 46°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
Linear Formula SnCl2·xH2O
UN Number UN3260
Merck Index 14,8783
Solubility Information Soluble in ethanol,acetone,ether and tetrahydrofuran. Insoluble in xylene.
IUPAC Name dichlorotin
Formula Weight 189.62 (Anhydrous)
Percent Purity 99.995%
Grade Puratronic
Odor Odorless
Sensitivity Air Sensitive
Density 2.71 g/mL
Assay (metals basis)
Chemical Name or Material Tin(II) chloride hydrate
Show More Show Less
Hazard Category H290-H302+H332-H314-H317-H335-H373
Hazard Statement GHS H Statement
H314-H318-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P234-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 231-868-0
RTECSNumber XP8850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only