missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) chloride dihydrate, Reagent Grade

Catalog No. AA1153622
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1153622 100 g
AA1153636 500 g
AA11536A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1153622 Supplier Thermo Scientific Chemicals Supplier No. 01153622

CAS: 10025-69-1 | SnCl2·2H2O

Stannous chloride is used as a sensitizing agent in preparing glass and plastic for metalizing. It serves as a potent reducing agent. Other uses of stannous chloride are tin electroplating baths, corrosion inhibitors, polymers, thermoplastic elastomers, soldering flux, antioxidant, tanning agent and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10025-69-1
Molecular Formula SnCl2·2H2O
MDL Number MFCD00149863
Synonym Stannous chloride dihydrate

Specifications

Melting Point 43°C to 46°C
Physical Form Crystalline
Quantity 100 g
Solubility Information Soluble in dilute and concentrated HCl,alcohol,ethyl acetate,glacial acetic acid,and NaOH solution. Decomposes in excess water.
Formula Weight 225.65 (189.62 Anhydrous)
Grade Reagent
Sensitivity Air Sensitive
Density 2.71 g/mL
Chemical Name or Material Tin(II) chloride dihydrate, Reagent Grade
Hazard Category H290-H314-H317-H332-H335-H373
Precautionary Statement P234-P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 600-045-1
RTECSNumber XP8850000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only