missing translation for 'onlineSavingsMsg'
Learn More

Tin(Ii) Chloride Dihydrate, puriss. p.a. ACS Reagent, Honeywell Fluka™

Catalog No. p-7072045
Change view
Click to view available options
Quantity:
500 g
1 kg
6 x 1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001697 500 g
6001696 1 kg
60045971 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6001697 Supplier Honeywell Chemicals Supplier No. 31669500G
Only null left

Puriss. p.a., ACS Reagent, Reagent ISO, Reagent European Pharmacopoeia,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10025-69-1
MDL Number MFCD00149863

Specifications

Quantity 500 g
Linear Formula SnCl2 · 2H2O
UN Number 3260
Formula Weight 225.65g/mol
Percent Purity ≥98%
Grade ACS/puriss. p.a.
Chemical Name or Material Tin(Ii) Chloride Dihydrate, Reag. Ph. Eur. reag. ISO
Hazard Statement P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H314-H317-H332-H335-H341-H361-H373-H410
EINECSNumber 231-868-0
RTECSNumber XP8850000