missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) chloride dihydrate, low in Mercury, suitable for AAS

Catalog No. p-7077809
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459600500 50 g
AC459602500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459600500 Supplier Thermo Scientific Chemicals Supplier No. 459600500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10025-69-1 | Cl2Sn·2H2O | 225.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10025-69-1
Molecular Formula Cl2Sn·2H2O
Molecular Weight (g/mol) 225.63
Synonym Stannous chloride dihydrate

Specifications

Boiling Point 652.0°C
Melting Point 37.0°C to 38.0°C
Quantity 50 g
Linear Formula SnCl2·2H2O
Solubility Information Solubility in water: 1187g/L (20°C). Other solubilities: soluble in alkalies and hcl, soluble in tartaric acid, alcohol, ethyl acetate, and glacial acetic acid
Formula Weight 225.63
Percent Purity ≥97.5% (Iodometry)
Chemical Name or Material Tin(II) chloride dihydrate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be corrosive to metals.
Harmful if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
May cause damage to organs through prolon
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER

RUO – Research Use Only