missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) Chloride Dihydrate, For Analysis, EMSURE™Test Kit, For water determination, According to Karl Fischer, Apura™, MilliporeSigma™

Catalog No. M1078150250
Change view
Click to view available options
Quantity:
100 g
250 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1078150250 250 g Glass Bottle
M1078150100 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1078150250 Supplier Technidata Supplier No. 1.07815.0250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10025-69-1
Molecular Formula SnCl2·2H2O
Synonym Tin Dichloride

Specifications

Boiling Point 623°C (1013hPa)(anhydrous substance)
Melting Point 38°C (anhydrous substance)
pH 1 to 2 (100g/L, H2O, 20°C)
Quantity 250 g
Solubility Information 1187g/L
Formula Weight 225.63
Percent Purity 98.0 to 103.0%
Grade ACS/ISO/Reagent Ph. Eur.
Packaging Glass Bottle
Density 2.71g/3 (20°C)
EINECSNumber 231-868-0