missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) chloride, anhydrous, 98%

Catalog No. AAA1620222
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1620222 100 g
AAA1620236 500 g
AAA162020E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1620222 Supplier Thermo Scientific Chemicals Supplier No. A1620222
Only null left

CAS: 7772-99-8 | Cl2Sn | 189.61 g/mol

Tin(II) chloride is widely used as a reducing agent in organic synthesis, for example conversion of quinones into hydroquinones, nitroaromatics into aromatic amines, and in Stephen reduction. It can be used to reduce certain metal salts into metals, tin plating, analytical testing of mercury ions, formation of silver mirrors and metal-metal bonds. It is also used for tin-plating, as a mordant in textile dyeing and as a catalyst in the production of polylactic acid. Further it is used as a food additive, wherein it serves as an anti-oxidant and a color retention agent. It can be used in the condensation of aryl aldehydes with cyclohexan-1,3-diones to synthesize xanthenes. It catalyzes direct conversion of aldehydes into beta-keto esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tin(II) chloride is widely used as a reducing agent in organic synthesis, for example conversion of quinones into hydroquinones, nitroaromatics into aromatic amines, and in Stephen reduction. It can be used to reduce certain metal salts into metals, tin plating, analytical testing of mercury ions, formation of silver mirrors and metal-metal bonds. It is also used for tin-plating, as a mordant in textile dyeing and as a catalyst in the production of polylactic acid. Further it is used as a food additive, wherein it serves as an anti-oxidant and a color retention agent. It can be used in the condensation of aryl aldehydes with cyclohexan-1,3-diones to synthesize xanthenes. It catalyzes direct conversion of aldehydes into beta-keto esters.

Solubility
Soluble in ethanol, acetone, ether and tetrahydrofuran. Insoluble in xylene.

Notes
Tin(II) chloride is very freely soluble in cold water. If the volume of water increases or at hot condition it hydrolyses to form an insoluble hydroxide derivative.

Chemical Identifiers

CAS 7772-99-8
Molecular Formula Cl2Sn
Molecular Weight (g/mol) 189.61
MDL Number MFCD00011241
InChI Key AXZWODMDQAVCJE-UHFFFAOYSA-L
Synonym Stannous chloride, anhydrous
PubChem CID 24479
ChEBI CHEBI:78067
IUPAC Name dichlorotin
SMILES [Cl-].[Cl-].[Sn++]

Specifications

Boiling Point 652°C
Melting Point 247°C
Quantity 100 g
UN Number UN3260
Merck Index 14,8783
Solubility Information Soluble in ethanol,acetone,ether and tetrahydrofuran. Insoluble in xylene.
Formula Weight 189.62
Percent Purity 98%
Sensitivity Air sensitive; Hygroscopic
Density 3.95
Chemical Name or Material Tin(II) chloride, anhydrous
Show More Show Less
Hazard Category H290-H302+H332-H314-H317-H335-H373
Hazard Statement GHS H Statement
H314-H318-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P234-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 231-868-0
RTECSNumber XP8700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only