missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) 2-Ethylhexanoate 85.0+%, TCI America™

Catalog No. p-7137605
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3149100G 100 g
T3149500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3149100G Supplier TCI America Supplier No. T3149100G
Only null left

Chemical Identifiers

CAS 301-10-0
Molecular Formula C16H30O4Sn
Molecular Weight (g/mol) 405.122
MDL Number MFCD00002676
InChI Key KSBAEPSJVUENNK-UHFFFAOYSA-L
Synonym stannous octoate, tin ii 2-ethylhexanoate, tin octoate, tin ii bis 2-ethylhexanoate, tin 2+ bis 2-ethylhexanoate, tin ethylhexanoate, tin 2-ethylhexanoate, hexanoic acid, 2-ethyl-, tin 2+ salt, stannous 2-ethylhexoate, tin ii 2-ethylhexylate
PubChem CID 9318
IUPAC Name 2-ethylhexanoate;tin(2+)
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Sn+2]

Specifications

Color Yellow
Quantity 100 g
Formula Weight 405.12
Percent Purity ≥85.0% (T)
Physical Form Liquid
Chemical Name or Material Tin(II) 2-Ethylhexanoate
EINECSNumber (2)-0615
RTECSNumber MO7870000
TSCA Yes