Learn More
Tin(II) 2-ethylhexanoate, 95%
CAS: 301-10-0 | C16H30O4Sn | 405.122 g/mol
$84.56 - $171.40
Chemical Identifiers
| CAS | 301-10-0 |
|---|---|
| Molecular Formula | C16H30O4Sn |
| Molecular Weight (g/mol) | 405.122 |
| MDL Number | MFCD00002676 |
| InChI Key | KSBAEPSJVUENNK-UHFFFAOYSA-L |
| Synonym | stannous octoate, tin ii 2-ethylhexanoate, tin octoate, tin ii bis 2-ethylhexanoate, tin 2+ bis 2-ethylhexanoate, tin ethylhexanoate, tin 2-ethylhexanoate, hexanoic acid, 2-ethyl-, tin 2+ salt, stannous 2-ethylhexoate, tin ii 2-ethylhexylate |
| PubChem CID | 9318 |
| IUPAC Name | 2-ethylhexanoate;tin(2+) |
| SMILES | CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Sn+2] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2361214
|
Thermo Scientific Chemicals
B2361214 |
25 g |
Each for $84.56
|
|
|||||
|
AAB2361222
|
Thermo Scientific Chemicals
B2361222 |
100 g |
Each for $171.40
|
|
|||||
Description
Tin(II) 2-ethylhexanoate is used as a polymerization initiator in polylactic acid production. It acts as an intermediate as well as a catalyst for urethane foams, a lubricant, addition agent and a stabilizer for transformer oils.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTin(II) 2-ethylhexanoate is used as a polymerization initiator in polylactic acid production. It acts as an intermediate as well as a catalyst for urethane foams, a lubricant, addition agent and a stabilizer for transformer oils.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
| 301-10-0 | |
| 405.122 | |
| KSBAEPSJVUENNK-UHFFFAOYSA-L | |
| 9318 | |
| CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Sn+2] |
| C16H30O4Sn | |
| MFCD00002676 | |
| stannous octoate, tin ii 2-ethylhexanoate, tin octoate, tin ii bis 2-ethylhexanoate, tin 2+ bis 2-ethylhexanoate, tin ethylhexanoate, tin 2-ethylhexanoate, hexanoic acid, 2-ethyl-, tin 2+ salt, stannous 2-ethylhexoate, tin ii 2-ethylhexylate | |
| 2-ethylhexanoate;tin(2+) |
Specifications
| 301-10-0 | |
| >110°C (230°F) | |
| C16H30O4Sn | |
| MFCD00002676 | |
| stannous octoate, tin ii 2-ethylhexanoate, tin octoate, tin ii bis 2-ethylhexanoate, tin 2+ bis 2-ethylhexanoate, tin ethylhexanoate, tin 2-ethylhexanoate, hexanoic acid, 2-ethyl-, tin 2+ salt, stannous 2-ethylhexoate, tin ii 2-ethylhexylate | |
| KSBAEPSJVUENNK-UHFFFAOYSA-L | |
| 2-ethylhexanoate;tin(2+) | |
| 9318 | |
| 96% |
| 1.251 | |
| Characteristic | |
| 1.494 | |
| 25 g | |
| Miscible with water. | |
| CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Sn+2] | |
| 405.122 | |
| 405.11 | |
| Tin(II) 2-ethylhexanoate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P272-P280g-P281-P302+P352-P305+P351+P338-P308+P313-P310-P333+P313-P363-P501c
H317-H318-H361
EINECSNumber : 206-108-6
RTECSNumber : MO7870000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only