missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin granules, ACS, 99.5% min
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Gray |
| Appearance | Gray granules |
| Quantity | 100 g |
| Odor | Odorless |
| Form | Granular |
| Assay Percent Range | 99.9% (Metals basis) |
| Grade | ACS |
| Chemical Name or Material | Tin, For analysis ACS |
Safety and Handling
| Hazard Category | H303: May be harmful if swallowed. |
| EINECSNumber | 231-141-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only