missing translation for 'onlineSavingsMsg'
Learn More

Tin Granulated, For analysis (Particle Size about 4mm) Reag. Ph Eur, EMSURE™, MilliporeSigma™

Catalog No. M1078061000
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1078061000 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1078061000 Supplier Technidata Supplier No. 1.07806.1000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Appearance Silver solid
Quantity 1 kg
Odor Odorless
Packaging Plastic bottle
Form Solid
Formula Weight 118.71
Chemical Name or Material Tin