missing translation for 'onlineSavingsMsg'
Learn More

Tin Foil, About 0.04mm Thick, MilliporeSigma™

Catalog No. M1078260001
Change view
Click to view available options
Quantity:
200 Strips
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1078260001 200 Strips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1078260001 Supplier Technidata Supplier No. 1.07826.0001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Appearance Silver solid
Quantity 200 Strips
Odor Odorless
Packaging Plastic box
Form Solid
Formula Weight 118.71. (molar mass)
Chemical Name or Material Tin