missing translation for 'onlineSavingsMsg'
Learn More

Tin foil, 0.25mm (0.01in) thick, Puratronic™, 99.998% (metals basis)

Catalog No. AA11459FI
Change view
Click to view available options
Quantity:
50 x 50 mm
100 x 100 mm
100 x 500 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA11459FI 50 x 50 mm
AA11459GH 100 x 100 mm
AA11459HP 100 x 500 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA11459FI Supplier Thermo Scientific Chemicals Supplier No. 011459FI

Form: approx. 4.6g/50x50mm | CAS: 7440-31-5 | Sn | 118.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Color Silver to Gray
Appearance Silver Gray foil
Density 7.31g/cm3
Quantity 50 x 50 mm
Odor Odorless
Form Foil
Assay Percent Range 99.998% (Metals basis)
Chemical Name or Material Tin
EINECSNumber 231-141-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only