missing translation for 'onlineSavingsMsg'
Learn More

Tin foil, 1.0mm (0.04in) thick, Puratronic™, 99.9985% (metals basis)

Form: Approx. 9.1g/25 x 50mm | CAS: 7440-31-5 | Sn | 118.71 g/mol

$146.88 - $1475.79

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA11457FG
View Documents
Thermo Scientific Chemicals
011457FG
25 x 50 mm
Each for $146.88
Only null left
 
AA11457FY
View Documents
Thermo Scientific Chemicals
011457FY
50 x 100 mm
Each for $388.37
Only null left
 
AA11457HL
View Documents
Thermo Scientific Chemicals
011457HL
100 x 250 mm
Each for $1,475.79
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-31-5
118.71
ATJFFYVFTNAWJD-UHFFFAOYSA-N
5352426
tin
Sn
MFCD00133862
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
CHEBI:32990
[Sn]

Specifications

Silver to Gray
Silver Gray foil
25 x 50 mm
99.9985% (Metals basis)
Sn
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
[Sn]
118.71
CHEBI:32990
7440-31-5
7.31g/cm3
Odorless
Foil
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
tin
5352426
Tin
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-141-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only