missing translation for 'onlineSavingsMsg'
Learn More

Tin foil, 1.0mm (0.04in) thick, 99.8% (metals basis)

CAS: 7440-31-5 | Sn | 118.71 g/mol

$78.03 - $619.62

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA43235FI
View Documents
Thermo Scientific Chemicals
043235FI
50 x 50 mm N/A
Only null left
 
AA43235GH
View Documents
Thermo Scientific Chemicals
043235GH
100 x 100 mm N/A
Only null left
 
AA43235HP
View Documents
Thermo Scientific Chemicals
043235HP
100 x 500 mm N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-31-5
118.71
ATJFFYVFTNAWJD-UHFFFAOYSA-N
5352426
tin
Sn
MFCD00133862
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
CHEBI:32990
[Sn]

Specifications

7440-31-5
Odorless
Foil
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
tin
5352426
99.8%
(metals basis)
50 x 50 mm
≈18.2g/50x50mm
Sn
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
[Sn]
118.71
CHEBI:32990
Tin foil, 1.0mm (0.04 in.) thick
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-141-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only