missing translation for 'onlineSavingsMsg'
Learn More

Tin foil, 0.1mm (0.004in) thick, 98.8% (metals basis)

Catalog No. AA44154RG
Change view
Click to view available options
Quantity:
30 x 150 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA44154RG 30 x 150 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA44154RG Supplier Thermo Scientific Chemicals Supplier No. 044154RG

missing translation for 'validMainframeMsg'

CAS: 7440-31-5 | Sn | 118.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Quantity 30 x 150 cm
Odor Odorless
Weight ≈329g/30x150cm
Form Foil
Percent Purity 98.8%
Assay (metals basis)
Chemical Name or Material Tin foil, 0.1mm (0.004 in.) thick
EINECSNumber 231-141-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only