missing translation for 'onlineSavingsMsg'
Learn More

Tin foil, 0.05mm (0.002in) thick, 98.8% (metals basis)

Catalog No. p-7051588
Change view
Click to view available options
Quantity:
15 x 15 cm
15 x 75 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41604VA 15 x 15 cm
AA41604VE 15 x 75 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA41604VA Supplier Thermo Scientific Chemicals Supplier No. 041604VA
Only null left

CAS: 7440-31-5 | Sn | 118.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Quantity 15 x 15 cm
Odor Odorless
Weight ≈8.2g/15x15cm
Form Foil
Percent Purity 98.8%
Assay (metals basis)
Chemical Name or Material Tin foil, 0.05mm (0.002 in.) thick
EINECSNumber 231-141-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only