Learn More
Costech Analytical Technologies Inc Tin Capsules, for Solid Samples, 3.5x5 mm, 100 per Pack
Catalog No. NC9601237
Encompass
Tin capsules for solid samples,Size: 3.5x5 mm,Consumables for the ECS 4010(Elemental Combustion System),NA 1500 Series 1108,NA 2500 and EA 1110 Analyzers,Flash EA 1112,NA 1500 Series 1 Analyzer,EA 1106 Analyzer,Europa ANCA N/C A
Catalog No. NC9601237 Supplier Costech Analytical Technologies Inc Supplier No. 041060
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Tin capsules for solid samples,Size: 3.5x5 mm,Consumables for the ECS 4010(Elemental Combustion System),NA 1500 Series 1108,NA 2500 and EA 1110 Analyzers,Flash EA 1112,NA 1500 Series 1 Analyzer,EA 1106 Analyzer,Europa ANCA N/C A