missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ TIMEtoken Clip-on Badge
Description
- Low cost clip-on paper badges
- With time-depenedent "backs" printed on them
- Apply the TIMEtoken or TIMEspot badge over printed "back" to start expiration
- Badges expire in one day
Specifications
Specifications
| Length (English) | 3 in. |
| Color | Red on white |
| Dimensions (L x W) | 3 in. x 4 in. |
| For Use With (Application) | Contractor |
| Width (English) | 4 in. |