Learn More
Research Products Corporation Timentin [Ticarcillin Disodium Salt/Potassium Clavulanate Mixture 15:1 Ratio], 1 Gram
Catalog No. 501368640
Encompass
Timentin contains two antibiotics: ticarcillin (a penicillin) and clavulanate potassium (a beta-lactamase inhibitor). The combination of ticarcillin and clavulanate enhances the spectrum of antibacterial activity by protecting ticarcillin from degradation by certain bacterial enzymes. 
Ticarcillin inhibits bacterial cell wall synthesis, disrupting the integrity of the bacterial cell wall and leading to bacterial cell lysis. Clavulanate potassium works by inhibiting beta-lactamases, enzymes produced by bacteria that can inactivate beta-lactam antibiotics, such as ticarcillin. This combination is effective against a broad spectrum of bacteria, including those producing beta-lactamases. 
Catalog No. 501368640 Supplier Research Products Corporation Supplier No. T360001.0
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Timentin contains two antibiotics: ticarcillin (a penicillin) and clavulanate potassium (a beta-lactamase inhibitor). The combination of ticarcillin and clavulanate enhances the spectrum of antibacterial activity by protecting ticarcillin from degradation by certain bacterial enzymes. 
Ticarcillin inhibits bacterial cell wall synthesis, disrupting the integrity of the bacterial cell wall and leading to bacterial cell lysis. Clavulanate potassium works by inhibiting beta-lactamases, enzymes produced by bacteria that can inactivate beta-lactam antibiotics, such as ticarcillin. This combination is effective against a broad spectrum of bacteria, including those producing beta-lactamases.