Learn More
Research Products Corporation Time Tape, Orange, 1 Inch Core, 1 Inch Wide, 500 Inch Roll, 6 Rolls per Case
Catalog No. 50998694
Encompass
Labeling tapes suited for a wide variety of laboratory uses. Easy to write on with pencil, pen, or permanent marker. Strong, durable tapes are resistant to oil, water, and acids. Withstand extreme temperature ranges from -10°F up to +250°F. Can be applied at temperatures as low as +32°F. Features a removable adhesive that peels off glass, plastic or metal.
Catalog No. 50998694 Supplier Research Products Corporation Supplier No. RPI 512502O
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Labeling tapes suited for a wide variety of laboratory uses. Easy to write on with pencil, pen, or permanent marker. Strong, durable tapes are resistant to oil, water, and acids. Withstand extreme temperature ranges from -10°F up to +250°F. Can be applied at temperatures as low as +32°F. Features a removable adhesive that peels off glass, plastic or metal.