Learn More
Fisherbrand™ Tilting Ergonomic Binocular Head
Catalog No. 03000118
Change view
Click to view available options
For Use With (Equipment):
Microscope
Catalog No. For Use With (Equipment)
03000118 Microscope
1 options
Catalog No. 03000118 Supplier Fisherbrand™ Supplier No. LMCPUTBI
Tilting ergonomic binocular head for upright compound microscope.
  • Ergonomic infinity gemel binocular head for LMC2P.
  • 30° to 60° elevation adjustable.
  • 360° rotatable.
  • Interpupillary adjustable distance: 54 to 75 mm.
Specifications
Type Binocular Head
For Use With (Equipment) Microscope