missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tigloyl Chloride 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 145°C |
| Quantity | 5 g |
| UN Number | 2920 |
| Formula Weight | 118.56 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | Tigloyl Chloride |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |