missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tiglic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 64°C |
| Boiling Point | 82°C |
| Color | White |
| UN Number | 3261 |
| Formula Weight | 100.12 |
| Percent Purity | ≥98.0% (GC,T) |
| Quantity | 500 g |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Tiglic Acid |
Safety and Handling
| EINECSNumber | (2)-0609&(2)-0968 |
| RTECSNumber | GQ5430000 |
| TSCA | Yes |
| Recommended Storage | Refrigerator |