missing translation for 'onlineSavingsMsg'
Learn More

Tiglic Acid 98.0+%, TCI America™

Catalog No. t0246100g
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0246100G 100 g
T024625G 25 g
T0246500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0246100G Supplier TCI America Supplier No. T0246100G
Only null left

Chemical Identifiers

CAS 80-59-1
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00066864
InChI Key UIERETOOQGIECD-ONEGZZNKSA-N
Synonym tiglic acid, tiglinic acid, cevadic acid, 2-methyl-2-butenoic acid, trans-2-methylcrotonic acid, trans-2,3-dimethylacrylic acid, 2-methylbut-2-enoic acid, e-2-methyl-2-butenoic acid, trans-2-methyl-2-butenoic acid, e-2,3-dimethylacrylic acid
PubChem CID 125468
ChEBI CHEBI:9592
IUPAC Name (2E)-2-methylbut-2-enoic acid
SMILES C\C=C(/C)C(O)=O

Specifications

Melting Point 64°C
Boiling Point 82°C
Color White
UN Number 3261
Formula Weight 100.12
Percent Purity ≥98.0% (GC,T)
Quantity 100 g
Physical Form Crystalline Powder
Chemical Name or Material Tiglic Acid
EINECSNumber (2)-0609&(2)-0968
RTECSNumber GQ5430000
TSCA Yes
Recommended Storage Refrigerator