missing translation for 'onlineSavingsMsg'
Learn More

Tiglic acid, 97+%

CAS: 80-59-1 | C5H8O2 | 100.12 g/mol

$85.26 - $748.49

Chemical Identifiers

CAS 80-59-1
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00066864
InChI Key UIERETOOQGIECD-ONEGZZNKSA-N
Synonym tiglic acid, tiglinic acid, cevadic acid, 2-methyl-2-butenoic acid, trans-2-methylcrotonic acid, trans-2,3-dimethylacrylic acid, 2-methylbut-2-enoic acid, e-2-methyl-2-butenoic acid, trans-2-methyl-2-butenoic acid, e-2,3-dimethylacrylic acid
PubChem CID 125468
ChEBI CHEBI:9592
IUPAC Name (E)-2-methylbut-2-enoic acid
SMILES C\C=C(/C)C(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1390514
View Documents
Thermo Scientific Chemicals
A1390514
25 g
Each for $85.26
Only null left
 
AAA1390522
View Documents
Thermo Scientific Chemicals
A1390522
100 g
Each for $160.64
Only null left
 
AAA1390536
View Documents
Thermo Scientific Chemicals
A1390536
500 g
Each for $748.49
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

80-59-1
100.12
UIERETOOQGIECD-ONEGZZNKSA-N
125468
(E)-2-methylbut-2-enoic acid
C5H8O2
MFCD00066864
tiglic acid, tiglinic acid, cevadic acid, 2-methyl-2-butenoic acid, trans-2-methylcrotonic acid, trans-2,3-dimethylacrylic acid, 2-methylbut-2-enoic acid, e-2-methyl-2-butenoic acid, trans-2-methyl-2-butenoic acid, e-2,3-dimethylacrylic acid
CHEBI:9592
C\C=C(/C)C(O)=O

Specifications

62°C to 66°C
198°C to 199°C
MFCD00066864
1236500
tiglic acid, tiglinic acid, cevadic acid, 2-methyl-2-butenoic acid, trans-2-methylcrotonic acid, trans-2,3-dimethylacrylic acid, 2-methylbut-2-enoic acid, e-2-methyl-2-butenoic acid, trans-2-methyl-2-butenoic acid, e-2,3-dimethylacrylic acid
C\C=C(/C)C(O)=O
100.12
CHEBI:9592
≥98%
25 g
Tiglic acid
80-59-1
C5H8O2
UN3261
149433
UIERETOOQGIECD-ONEGZZNKSA-N
(E)-2-methylbut-2-enoic acid
125468
100.12
0.969
101°C (213°F)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 201-295-0

RTECSNumber : GQ5430000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only