missing translation for 'onlineSavingsMsg'
Learn More

Tiglic acid, 97+%

Catalog No. aaa1390522
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1390522 100 g
AAA1390514 25 g
AAA1390536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1390522 Supplier Thermo Scientific Chemicals Supplier No. A1390522
Only null left

CAS: 80-59-1 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80-59-1
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00066864
InChI Key UIERETOOQGIECD-ONEGZZNKSA-N
Synonym tiglic acid, tiglinic acid, cevadic acid, 2-methyl-2-butenoic acid, trans-2-methylcrotonic acid, trans-2,3-dimethylacrylic acid, 2-methylbut-2-enoic acid, e-2-methyl-2-butenoic acid, trans-2-methyl-2-butenoic acid, e-2,3-dimethylacrylic acid
PubChem CID 125468
ChEBI CHEBI:9592
IUPAC Name (E)-2-methylbut-2-enoic acid
SMILES C\C=C(/C)C(O)=O

Specifications

Melting Point 62°C to 66°C
Boiling Point 198°C to 199°C
UN Number UN3261
Beilstein 1236500
Merck Index 149433
Formula Weight 100.12
Percent Purity ≥98%
Density 0.969
Quantity 100 g
Flash Point 101°C (213°F)
Chemical Name or Material Tiglic acid
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 201-295-0
RTECSNumber GQ5430000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only