missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl Orthoformate, 99%, Spectrum™ Chemical

Catalog No. 18614566
Change view
Click to view available options
Quantity:
500 g
200 L
Packaging:
Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614566 500 g Drum
18614565 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18614566 Supplier Spectrum Chemical Mfg Cor Supplier No. TR124500GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 149-73-5

Specifications

CAS Max % 1
Boiling Point 100.6°C to 102°C (213.1°F to 216°F
Packaging Drum
Refractive Index 1.3765-1.3815
Quantity 500 g
Molecular Weight (g/mol) 106.12
Percent Purity 0.99
Grade Ungraded
Physical Form Liquid
Chemical Name or Material Trimethyl Orthoformate

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.