Learn More
Eisco™ Ticker Tape Timer
Description
- Housed in moulded plastic case
- Timer operates on 12 volts AC supply, Power supply is not included
- With tape guide and moveable carbon disc plug to maximise use of discs
- Supplied with one roll of 50 meter paper tape and carbon discs box of 100
Specifications
Specifications
| Product Type | Ticker Tape Timer |
| Description | Operates on 12 volts AC supply (Power supply is not included) |
| Material | Molded Plastic Case |
| For Use With (Application) | Produces 50/60 dots per second depending on main frequency |
| Includes | One roll of 50 meter paper tape and carbon discs box of 100 |