missing translation for 'onlineSavingsMsg'
Learn More

Ticarcillin Impurity C, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022852
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022852 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022852 Supplier Merck Emd Millipore Supplier No. BP1080

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

CAS 21080-92-2
Melting Point 139°C (decomposition) (literature)
Molecular Formula C7H6O4S
Linear Formula C7H6O4S
Quantity 25 mg
MDL Number MFCD00005469
Synonym 3-Thiophenemalonic acid; alpha-Carboxythiophene-3-acetic acid; (3-Thienyl)malonic acid
Formula Weight 186.19
Grade BP