Learn More
Tians High Performance SMS Lab Coats
Catalog No. 191501558
Change view
Click to view available options
Size:
Small
Medium
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
191501558 -
191501554 Medium
191501554A Small
191501557 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 191501558 Supplier Tians Supplier No. 864795 3XL
Low linting lab coats for use in critical and low particulate clean environments.
  • Constructed of SMS fabric that is strong and durable, yet lightweight and resistant to tears and puncture
  • Features knit collar, left chest pocket and two side pockets with snap front and knit wrist
  • Provides fluid and particulate safety barrier without sacrificing mobility and comfort
  • Impede the spread of viruses, germs, impurities and infections
Specifications
Product Type Controlled Environments Lab Coat
Material SMS
Color Blue
Closure Type Snaps
Cuff Style Knit
Certifications/Compliance ASTM D751, ASTM D3787, ASTM D2582, ASTM E96-80
Item Description White Wrists, White Collar
Packaging Type Neatly Folded, Cleanroom Packed
Cleanroom Class Class 1000 (ISO 6)