Learn More
Tians High Performance SMS Lab Coats
Description
- Constructed of SMS fabric that is strong and durable, yet lightweight and resistant to tears and puncture
- Features knit collar, left chest pocket and two side pockets with snap front and knit wrist
- Provides fluid and particulate safety barrier without sacrificing mobility and comfort
- Impede the spread of viruses, germs, impurities and infections
Specifications
Specifications
| Product Type | Controlled Environments Lab Coat |
| Color | Blue |
| Closure Type | Snaps |
| Cuff Style | Knit |
| Item Description | White Wrists, White Collar |
| Packaging Type | Neatly Folded, Cleanroom Packed |
| Cleanroom Class | Class 1000 (ISO 6) |