missing translation for 'onlineSavingsMsg'
Learn More

Tians epic™ High Performance Static Dissipative Cleanroom Frocks, Knit Wrists and Collar

Catalog No. 19161130G
Change view
Click to view available options
Size (English) Chest:
11 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (English) Chest
19161130G 11 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19161130G Supplier Tians Supplier No. 846855 5XL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Lint-free film coated frock for use in critical, ESD, and low particulate clean environments.

  • Lint-free film coated frock
  • Suitable for exposure to bloodborne pathogens, some hazardous liquids, and any wet general maintenance areas
  • Features include a snap front, knit wrists
  • Knit collar, no pockets
  • Fabric is constructed from a blend of polyethylene and polypropylene
  • Finished fabric contains a proprietary additive making it static dissipative
  • For use in ISO 6 Cleanroom Class environments
  • Neatly folded and cleanroom packed
  • 10 pieces per sealed inner polybag, 3 polybags per sealed master bag, 30 pieces per case

Specifications

Size 5X-Large
Material Polypropylene, Polyethylene
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Snaps
Cuff Style Knit
Collar Type Knit
Size (English) Chest 11 in.
Size (Metric) Chest 27.9 cm
Certifications/Compliance Class 1 CPSC 1610, IEST-RP-CC003.3
Packaging Type Cleanroom Packed
Dimensions (L x W) 17 x 11 in. (43.1 x 27.9 cm)
Length (Metric) 43.1 cm
Cleanroom Class Class 1000 (ISO 6)
Length (English) 17 in.
Product Type Controlled Environments Frock
Show More Show Less