missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Tiamulin fumarate, 95%
CAS: 55297-96-6 | C32H51NO8S | 609.82 g/mol
Supplier: Thermo Scientific™ J66020ME
Description
Targets Gram-positive bacteria as well as a few mycoplasma species
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Targets Gram-positive bacteria as well as a few mycoplasma speciesSpecifications
| 55297-96-6 | |
| 563° | |
| C32H51NO8S | |
| 500 mg | |
| Thiamutilin fumarate; | |
| OC(=O)\C=C\C(O)=O.CCN(CC)CCSCC(=O)O[C@@H]1C[C@@](C)(C=C)[C@@H](O)[C@H](C)C23CCC(=O)[C@@H]2C1(C)[C@H](C)CC3 | |
| 609.82 | |
| 95% |
| 147-148° | |
| >110°(230°F) | |
| MFCD00145407 | |
| 14,9419 | |
| YXQXDXAHCSEVSD-LKNSULBJSA-N | |
| (2E)-but-2-enedioic acid; (2R,3S,4S,6R,8S,14R)-4-ethenyl-3-hydroxy-2,4,7,14-tetramethyl-9-oxotricyclo[5.4.3.0¹,â¸]tetradecan-6-yl 2-{[2-(diethylamino)ethyl]sulfanyl}acetate | |
| 609.81 | |
| Solid |
Chemical Identifiers
| 55297-96-6 | |
| 609.82 | |
| YXQXDXAHCSEVSD-LKNSULBJSA-N | |
| (2E)-but-2-enedioic acid; (2R,3S,4S,6R,8S,14R)-4-ethenyl-3-hydroxy-2,4,7,14-tetramethyl-9-oxotricyclo[5.4.3.0¹,â¸]tetradecan-6-yl 2-{[2-(diethylamino)ethyl]sulfanyl}acetate |
| C32H51NO8S | |
| MFCD00145407 | |
| Thiamutilin fumarate; | |
| OC(=O)\C=C\C(O)=O.CCN(CC)CCSCC(=O)O[C@@H]1C[C@@](C)(C=C)[C@@H](O)[C@H](C)C23CCC(=O)[C@@H]2C1(C)[C@H](C)CC3 |
Safety and Handling
ShelfLife : 5 Years
EINECSNumber : 259-581-6
RTECSNumber : AG9560000