Learn More
StatLab Thymus IHC Control Slides
Catalog No. 22024942
Change view
Click to view available options
Quantity:
25 Pk.
Catalog No. Quantity
22024942 25 Pk.
1 options
Catalog No. 22024942 Supplier StatLab Supplier No. CST0725P
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Use thymus IHC control slides with confidence to evaluate stain quality and performance in histology laboratories
  • Slides consist of formalin fixed paraffin embedded tissue sections which contain known distinctive tissue elements
  • Use these slides with confidence to evaluate stain quality and performance in histology laboratories
  • Includes 25 Unstained Slides per pack
Specifications
Product Type Control Slide
Quantity 25 Pk.