missing translation for 'onlineSavingsMsg'
Learn More

Thymoquinone

Catalog No. AC305070050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC305070050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC305070050 Supplier Thermo Scientific Chemicals Supplier No. 305070050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 490-91-5 | C10H12O2 | 164.20 g/mol

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 490-91-5
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
InChI Key KEQHJBNSCLWCAE-UHFFFAOYSA-N
IUPAC Name 2-methyl-5-(propan-2-yl)cyclohexa-2,5-diene-1,4-dione
SMILES CC(C)C1=CC(=O)C(C)=CC1=O

Specifications

Melting Point 43.0°C to 44.0°C
Boiling Point 230.0°C to 232.0°C
Flash Point 103°C
Quantity 5 g
Solubility Information Solubility aq. soln.: insoluble aq. soln.. Other solubilities: soluble in chloroform
Formula Weight 164.2
Chemical Name or Material Thymoquinone
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes serious eye irritation.

Harmful if swallowed.

May cause respiratory irritation.

Causes skin irritation.

Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

If skin irritation occurs: Get medical advice/attention.

If eye irritation persists: Get medical advice/attention.

Avoid breathing dust/fume/gas/mist/vapors/spray.

IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

EINECSNumber 207-721-1

RUO – Research Use Only