missing translation for 'onlineSavingsMsg'
Learn More

Thymoquinone 98.0+%, TCI America™

Catalog No. p-7136433
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T07951G 1 g
T07955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T07951G Supplier TCI America Supplier No. T07951G

Chemical Identifiers

CAS 490-91-5
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00001602
InChI Key KEQHJBNSCLWCAE-UHFFFAOYSA-N
Synonym thymoquinone, thymoquinon, p-cymene-2,5-dione, 2-isopropyl-5-methylbenzoquinone, polythymoquinone, 2,5-cyclohexadiene-1,4-dione, 2-methyl-5-1-methylethyl, 2-isopropyl-5-methyl-1,4-benzoquinone, 5-isopropyl-2-methyl-1,4-benzoquinone, 2-isopropyl-5-methyl-p-benzoquinone, p-mentha-3,6-diene-2,5-dione
PubChem CID 10281
IUPAC Name 2-methyl-5-propan-2-ylcyclohexa-2,5-diene-1,4-dione
SMILES CC1=CC(=O)C(=CC1=O)C(C)C

Specifications

Melting Point 46°C
Boiling Point 232°C
Quantity 1 g
Formula Weight 164.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Thymoquinone
RTECSNumber GU5330000
TSCA Yes