missing translation for 'onlineSavingsMsg'
Learn More

Thymoquinone, 99%

Catalog No. AC305070010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC305070010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC305070010 Supplier Thermo Scientific Chemicals Supplier No. 305070010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 490-91-5 | C10H12O2 | 164.204 g/mol

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 490-91-5
Melting Point 44°C to 47°C
Color Orange to Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C10H12O2
Quantity 1 g
Synonym thymoquinone, thymoquinon, p-cymene-2,5-dione, 2-isopropyl-5-methylbenzoquinone, polythymoquinone, 2,5-cyclohexadiene-1,4-dione, 2-methyl-5-1-methylethyl, 2-isopropyl-5-methyl-1,4-benzoquinone, 5-isopropyl-2-methyl-1,4-benzoquinone, 2-isopropyl-5-methyl-p-benzoquinone, p-mentha-3,6-diene-2,5-dione
InChI Key KEQHJBNSCLWCAE-UHFFFAOYSA-N
SMILES CC1=CC(=O)C(=CC1=O)C(C)C
IUPAC Name 2-methyl-5-propan-2-ylcyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol) 164.204
PubChem CID 10281
Formula Weight 164.2
Percent Purity ≥98.5%
Physical Form Crystals
Chemical Name or Material Thymoquinone, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only