missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thymolphthalexone

Catalog No. AAL1096203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1096203 1 g
AAL1096206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1096203 Supplier Thermo Scientific Chemicals Supplier No. L1096203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1913-93-5 | C38H44N2O12 | 720.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1913-93-5
Molecular Formula C38H44N2O12
Molecular Weight (g/mol) 720.77
MDL Number MFCD00001619
InChI Key AGXCADFSYJNMEG-UHFFFAOYNA-N
Synonym thymolphthalexone, thymolphthalein complexon, thymolphthalein complexone, thymolphthalexon, thymolphthalein-3,3'-bismethyliminodiacetic acid, 3-1-3-bis carboxymethyl amino methyl-4-hydroxy-5-isopropyl-2-methylphenyl-3-oxo-2-benzofuran-1-yl-6-hydroxy-5-isopropyl-2-methylphenyl methyl carboxymethyl amino acetic acid, n,n'-3-oxo-1 3h-isobenzofuranylidene bis 6-hydroxy-2-methyl-5-1-methylethyl-3,1-phenylene methylene bis n-carboxymethyl glycine, thymophthalexon, 3',3-bis n,n-di carboxymethyl aminomethyl thymolphthalein, 2,2',2,2'-3-oxo-1,3-dihydro-2-benzofuran-1,1-diyl bis 6-hydroxy-5-isopropyl-2-methyl-3,1-phenylene methylenenitrilo tetraacetic acid
PubChem CID 112918
IUPAC Name 2-[[5-[1-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-3-oxo-2-benzofuran-1-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid
SMILES CC(C)C1=CC(=C(C)C(C[NH+](CC(O)=O)CC([O-])=O)=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C(O)C(C[NH+](CC(O)=O)CC([O-])=O)=C1C

Specifications

Quantity 1 g
Beilstein 1339087
Formula Weight 720.78
Chemical Name or Material Thymolphthalexone
EINECSNumber 217-627-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only