missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thymolphthalein, pure, indicator

Catalog No. p-3734589
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC151460100 10 g Glass bottle
AC151460500 50 g Glass bottle
AC151462500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC151460100 Supplier Thermo Scientific Chemicals Supplier No. 151460100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 125-20-2 | C28H30O4 | 430.544 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 125-20-2
Molecular Formula C28H30O4
Molecular Weight (g/mol) 430.544
MDL Number MFCD00005909
InChI Key LDKDGDIWEUUXSH-UHFFFAOYSA-N
Synonym TP
PubChem CID 31316
IUPAC Name 3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one
SMILES CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O

Specifications

Melting Point 251.0°C to 253.0°C
Infrared Spectrum Authentic
Loss on Drying 1% max.
Packaging Glass bottle
Visual Transition Interval from pH 8.8 (colorless) to pH 10.5 (blue)
Beilstein 18, I, 381
Merck Index 15, 9556
Color White
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dilute alkali and acids, soluble in alcohol and acetone
Quantity 10 g
Physical Form Fine Crystalline Powder
Formula Weight 430.53
Grade Pure
Chemical Name or Material Thymolphthalein, Indicator
Show More Show Less

RUO – Research Use Only