missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thymolphthalein Indicator, 0.05% (w/v) in 90% (v/v) Alcohol, Ricca Chemical
$130.63 - $165.70
Specifications
| Chemical Name or Material | Thymolphthalein Indicator |
|---|---|
| CAS | 64-17-5,7732-18-5,67-56-1,125-20-2 |
| CAS Min % | 9.0,84.0,0.03,4.0 |
| CAS Max % | 11.0,88.0,0.03,6.0 |
| Concentration | 0.05% (w/v) |
Specifications
| Thymolphthalein Indicator | |
| 9.0,84.0,0.03,4.0 | |
| 0.05% (w/v) | |
| Indicator | |
| 90% (v/v) Alcohol |
| 64-17-5,7732-18-5,67-56-1,125-20-2 | |
| 11.0,88.0,0.03,6.0 | |
| 0.05% W/V | |
| Natural Poly Bottle | |
| Colorless |