missing translation for 'onlineSavingsMsg'
Learn More

Thymolphthalein, ACS Reagent Grade, Ricca Chemical

Catalog No. p-3339901
Change view
Click to view available options
Quantity:
5 g
Packaging:
Pouch
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDCT04505F1 5 g Pouch
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCT04505F1 Supplier Ricca Chemical Company Supplier No. RDCT04505F1
Only null left

Chemical Identifiers

CAS 125-20-2
Molecular Formula C28H30O4
Molecular Weight (g/mol) 430.544
InChI Key LDKDGDIWEUUXSH-UHFFFAOYSA-N
PubChem CID 31316
IUPAC Name 3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one
SMILES CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Grade ACS
Packaging Pouch
Color Colorless to White
Quantity 5 g
Chemical Name or Material Thymolphthalein
ShelfLife 60 months