missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thymolphthalein, ACS reagent

CAS: 125-20-2 | C28H30O4 | 430.53 g/mol

Supplier:  Thermo Scientific Chemicals 420955000

 View more versions of this product

Catalog No. AC420955000


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

Thymolphthalein
251.0°C to 253.0°C
Authentic
C28H30O4
18,I,381
thymolphthalein, thymophthalein, unii-yg5i28wsqp, yg5i28wsqp, phenolphthalein, 5',5-diisopropyl-2',2-dimethyl, 3,3-bis 4-hydroxy-5-isopropyl-2-methylphenyl-2-benzofuran-1 3h-one, 1 3h-isobenzofuranone, 3,3-bis 4-hydroxy-2-methyl-5-1-methylethyl phenyl, 5',5-diisopropyl-2',2-dimethylphenolphthalein, 3,3-bis 4-hydroxy-2-methyl-5-propan-2-ylphenyl-2-benzofuran-1-one
LDKDGDIWEUUXSH-UHFFFAOYSA-N
3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one
31316
passes test
Low Melting Mass
125-20-2
White to Yellow
Glass bottle
500 g
15,9556
Solubility in water: insoluble. Other solubilities: soluble in dilute alkali and acids, soluble in alcohol and acetone
CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O
430.53
430.53
ACS Reagent

Chemical Identifiers

125-20-2
430.53
thymolphthalein, thymophthalein, unii-yg5i28wsqp, yg5i28wsqp, phenolphthalein, 5',5-diisopropyl-2',2-dimethyl, 3,3-bis 4-hydroxy-5-isopropyl-2-methylphenyl-2-benzofuran-1 3h-one, 1 3h-isobenzofuranone, 3,3-bis 4-hydroxy-2-methyl-5-1-methylethyl phenyl, 5',5-diisopropyl-2',2-dimethylphenolphthalein, 3,3-bis 4-hydroxy-2-methyl-5-propan-2-ylphenyl-2-benzofuran-1-one
3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one
C28H30O4
LDKDGDIWEUUXSH-UHFFFAOYSA-N
31316
CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-729-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only