missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thymolphthalein, ACS reagent

Catalog No. AC420950100
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC420950100 10 g Glass bottle
AC420950250 25 g Glass bottle
AC420951000 100 g Glass bottle
AC420955000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC420950100 Supplier Thermo Scientific Chemicals Supplier No. 42095 0100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 125-20-2 | C28H30O4 | 430.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 125-20-2
Molecular Formula C28H30O4
Molecular Weight (g/mol) 430.53
InChI Key LDKDGDIWEUUXSH-UHFFFAOYSA-N
Synonym thymolphthalein, thymophthalein, unii-yg5i28wsqp, yg5i28wsqp, phenolphthalein, 5',5-diisopropyl-2',2-dimethyl, 3,3-bis 4-hydroxy-5-isopropyl-2-methylphenyl-2-benzofuran-1 3h-one, 1 3h-isobenzofuranone, 3,3-bis 4-hydroxy-2-methyl-5-1-methylethyl phenyl, 5',5-diisopropyl-2',2-dimethylphenolphthalein, 3,3-bis 4-hydroxy-2-methyl-5-propan-2-ylphenyl-2-benzofuran-1-one
PubChem CID 31316
IUPAC Name 3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one
SMILES CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O

Specifications

Melting Point 251.0°C to 253.0°C
Color White to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 10 g
Beilstein 18,I,381
Merck Index 15,9556
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dilute alkali and acids, soluble in alcohol and acetone
Formula Weight 430.53
Clarity passes test
Grade ACS Reagent
Physical Form Low Melting Mass
Chemical Name or Material Thymolphthalein
Show More Show Less
EINECSNumber 204-729-7

RUO – Research Use Only