missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thymolphthalein

CAS: 125-20-2 | C28H30O4 | 430.544 g/mol

Supplier:  Thermo Scientific Chemicals B2389609

 View more versions of this product

Catalog No. AAB2389609


Only null left

Description

Description

Thymolphthalein acts as pH indicator, as reagent for blood after decolorizing the alkaline solution by boiling with zinc dust.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thymolphthalein acts as pH indicator, as reagent for blood after decolorizing the alkaline solution by boiling with zinc dust.

Solubility
Soluble in alcohol, acetone and also soluble in dilute alkalis. insoluble in water.

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
Specifications

Specifications

Thymolphthalein
251°C to 253°C
MFCD00005909
359413
thymolphthalein, thymophthalein, unii-yg5i28wsqp, yg5i28wsqp, phenolphthalein, 5',5-diisopropyl-2',2-dimethyl, 3,3-bis 4-hydroxy-5-isopropyl-2-methylphenyl-2-benzofuran-1 3h-one, 1 3h-isobenzofuranone, 3,3-bis 4-hydroxy-2-methyl-5-1-methylethyl phenyl, 5',5-diisopropyl-2',2-dimethylphenolphthalein, 3,3-bis 4-hydroxy-2-methyl-5-propan-2-ylphenyl-2-benzofuran-1-one
LDKDGDIWEUUXSH-UHFFFAOYSA-N
3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one
31316
125-20-2
C28H30O4
10 g
14,9401
Soluble in alcohol,acetone and also soluble in dilute alkalis. insoluble in water.
CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O
430.544
430.55

Chemical Identifiers

125-20-2
430.544
LDKDGDIWEUUXSH-UHFFFAOYSA-N
31316
CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O
C28H30O4
MFCD00005909
thymolphthalein, thymophthalein, unii-yg5i28wsqp, yg5i28wsqp, phenolphthalein, 5',5-diisopropyl-2',2-dimethyl, 3,3-bis 4-hydroxy-5-isopropyl-2-methylphenyl-2-benzofuran-1 3h-one, 1 3h-isobenzofuranone, 3,3-bis 4-hydroxy-2-methyl-5-1-methylethyl phenyl, 5',5-diisopropyl-2',2-dimethylphenolphthalein, 3,3-bis 4-hydroxy-2-methyl-5-propan-2-ylphenyl-2-benzofuran-1-one
3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H500

EINECSNumber : 204-729-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only