missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thymol 99.0+%, TCI America™
$78.93 - $232.95
Chemical Identifiers
| CAS | 89-83-8 |
|---|---|
| Molecular Formula | C10H14O |
| Molecular Weight (g/mol) | 150.221 |
| MDL Number | MFCD00002309 |
| InChI Key | MGSRCZKZVOBKFT-UHFFFAOYSA-N |
| Synonym | thymol, 2-isopropyl-5-methylphenol, 5-methyl-2-isopropylphenol, thyme camphor, 3-p-cymenol, 6-isopropyl-m-cresol, thymic acid, isopropyl cresol, 5-methyl-2-1-methylethyl phenol, 6-isopropyl-3-methylphenol |
| PubChem CID | 6989 |
| ChEBI | CHEBI:27607 |
| IUPAC Name | 5-methyl-2-propan-2-ylphenol |
| SMILES | CC1=CC(=C(C=C1)C(C)C)O |
Chemical Identifiers
| 89-83-8 | |
| 150.221 | |
| MGSRCZKZVOBKFT-UHFFFAOYSA-N | |
| 6989 | |
| 5-methyl-2-propan-2-ylphenol |
| C10H14O | |
| MFCD00002309 | |
| thymol, 2-isopropyl-5-methylphenol, 5-methyl-2-isopropylphenol, thyme camphor, 3-p-cymenol, 6-isopropyl-m-cresol, thymic acid, isopropyl cresol, 5-methyl-2-1-methylethyl phenol, 6-isopropyl-3-methylphenol | |
| CHEBI:27607 | |
| CC1=CC(=C(C=C1)C(C)C)O |
Specifications
| 89-83-8 | |
| White | |
| C10H14O | |
| 25 g | |
| thymol, 2-isopropyl-5-methylphenol, 5-methyl-2-isopropylphenol, thyme camphor, 3-p-cymenol, 6-isopropyl-m-cresol, thymic acid, isopropyl cresol, 5-methyl-2-1-methylethyl phenol, 6-isopropyl-3-methylphenol | |
| CC1=CC(=C(C=C1)C(C)C)O | |
| 150.221 | |
| CHEBI:27607 | |
| ≥99.0% (GC) | |
| Thymol |
| 51°C | |
| 232°C | |
| MFCD00002309 | |
| 1759 | |
| MGSRCZKZVOBKFT-UHFFFAOYSA-N | |
| 5-methyl-2-propan-2-ylphenol | |
| 6989 | |
| 150.22 | |
| Crystalline Lumps |
Safety and Handling
EINECSNumber : (3)-0521&(4)-0057
RTECSNumber : XP2275000
TSCA : Yes