missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thymol Blue, TCI America™
$68.97 - $212.51
Chemical Identifiers
| CAS | 76-61-9 |
|---|---|
| Molecular Formula | C27H30O5S |
| Molecular Weight (g/mol) | 466.592 |
| MDL Number | MFCD00005869 |
| InChI Key | PRZSXZWFJHEZBJ-UHFFFAOYSA-N |
| Synonym | Thymolsulfonphthalein |
| PubChem CID | 65565 |
| IUPAC Name | 4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol |
| SMILES | CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O |
Chemical Identifiers
| 76-61-9 | |
| 466.592 | |
| PRZSXZWFJHEZBJ-UHFFFAOYSA-N | |
| 65565 | |
| CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O |
| C27H30O5S | |
| MFCD00005869 | |
| Thymolsulfonphthalein | |
| 4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol |
Specifications
| 222°C | |
| C27H30O5S | |
| Thymolsulfonphthalein | |
| CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O | |
| 466.592 | |
| 466.59 | |
| 1 g | |
| Thymol Blue |
| 76-61-9 | |
| MFCD00005869 | |
| PRZSXZWFJHEZBJ-UHFFFAOYSA-N | |
| 4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol | |
| 65565 | |
| Green/Yellow | |
| Crystal/Powder |
Safety and Handling
RTECSNumber : XP2575000
TSCA : Yes